Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001159554.1 | 2038 | Intron | NP_001153026.1 | ||
NM_005446.3 | 2038 | Intron | NP_005437.2 | ||
XM_005261819.3 | 2038 | Intron | XP_005261876.1 | ||
XM_011530498.2 | 2038 | Intron | XP_011528800.1 | ||
XM_011530499.2 | 2038 | Intron | XP_011528801.1 | ||
XM_011530500.2 | 2038 | Intron | XP_011528802.1 | ||
XM_011530501.2 | 2038 | Intron | XP_011528803.1 | ||
XM_011530502.2 | 2038 | Intron | XP_011528804.1 | ||
XM_017029066.1 | 2038 | Intron | XP_016884555.1 | ||
XM_017029067.1 | 2038 | Intron | XP_016884556.1 | ||
XM_017029068.1 | 2038 | Intron | XP_016884557.1 | ||
XM_017029069.1 | 2038 | Intron | XP_016884558.1 | ||
XM_017029070.1 | 2038 | Intron | XP_016884559.1 | ||
XM_017029071.1 | 2038 | Intron | XP_016884560.1 | ||
XM_017029072.1 | 2038 | Intron | XP_016884561.1 | ||
XM_017029073.1 | 2038 | Intron | XP_016884562.1 | ||
XM_017029074.1 | 2038 | Intron | XP_016884563.1 | ||
XM_017029075.1 | 2038 | Intron | XP_016884564.1 | ||
XM_017029076.1 | 2038 | Intron | XP_016884565.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_004173.2 | 2038 | UTR 3 | NP_004164.2 |