Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001111034.2 | 1064 | Missense Mutation | ATC,GTC | I221V | NP_001104504.1 |
NM_001111035.2 | 1064 | Missense Mutation | ATC,GTC | I221V | NP_001104505.1 |
NM_001111036.2 | 1064 | Missense Mutation | ATC,GTC | I221V | NP_001104506.1 |
NM_001322023.1 | 1064 | Missense Mutation | ATC,GTC | I221V | NP_001308952.1 |
NM_001611.4 | 1064 | Missense Mutation | ATC,GTC | I221V | NP_001602.1 |
XM_005259938.1 | 1064 | Missense Mutation | ATC,GTC | I221V | XP_005259995.1 |
XM_011528069.2 | 1064 | Missense Mutation | ATC,GTC | I221V | XP_011526371.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001290083.1 | 1064 | Intron | NP_001277012.1 | ||
NM_001290084.1 | 1064 | Intron | NP_001277013.1 | ||
NM_001290085.1 | 1064 | Intron | NP_001277014.1 | ||
NM_145295.3 | 1064 | Intron | NP_660338.1 | ||
XM_011527780.2 | 1064 | Intron | XP_011526082.1 |