Product Details
- SNP ID
-
rs2956112
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:34916354 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGCGCGCGGCGCTTAGCCTGGGAT[A/G]CGGCAGCGAGGCCGCAAATGCAATC
- Phenotype
-
MIM: 612491
MIM: 608769
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
APIP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113073242] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- APIP
- Gene Name
- APAF1 interacting protein
- Gene
- PDHX
- Gene Name
- pyruvate dehydrogenase complex component X
View Full Product Details