Product Details

SNP ID
rs2242225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:35953218 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTATCAGAGAGGTAGTTTGGTGTTA[C/T]ACTAAGGAAGATAAAAAAGAAAAGA
Phenotype
MIM: 616383
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UGT3A1 PubMed Links

Gene Details

Gene
UGT3A1
Gene Name
UDP glycosyltransferase family 3 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171873.1 2478 Intron NP_001165344.1
NM_152404.3 2478 UTR 3 NP_689617.3
XM_005248243.4 2478 UTR 3 XP_005248300.1
XM_011513957.2 2478 UTR 3 XP_011512259.1
XM_011513958.2 2478 UTR 3 XP_011512260.1
XM_011513959.2 2478 UTR 3 XP_011512261.1
XM_011513961.2 2478 UTR 3 XP_011512263.1

View Full Product Details