Product Details

SNP ID
rs2240229
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15742270 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCTTCCTCAGCCCAATCATTTTCA[A/G]TCTAAGGAACAAGGAGCTGAAGAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR10H3 PubMed Links

Gene Details

Gene
OR10H3
Gene Name
olfactory receptor family 10 subfamily H member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013938.1 878 Missense Mutation AAT,AGT N293S NP_039226.1

View Full Product Details