Product Details
- SNP ID
-
rs2287307
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:46124335 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- AGCAGACAAGAGCTAAGGGAGAAAG[A/G]AAAAAGGTTGAACATGGAGCTGAGA
- Phenotype
-
MIM: 610545
MIM: 610546
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IGFL2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10408134] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IGFL2
- Gene Name
- IGF like family member 2
- Gene
- IGFL3
- Gene Name
- IGF like family member 3
View Full Product Details