Product Details

SNP ID
rs2275154
Assay Type
Functionally tested
NCBI dbSNP Submissions
44
Location
Chr.1:243270352 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TCCATTCATTCTTCATTCTGCTGCT[A/G]TGATGTTATTCCTCAAACACAAATC
Phenotype
MIM: 613524
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SDCCAG8 PubMed Links

Gene Details

Gene
SDCCAG8
Gene Name
serologically defined colon cancer antigen 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006642.3 Intron NP_006633.1
XM_005273013.4 Intron XP_005273070.1
XM_005273018.1 Intron XP_005273075.1
XM_005273021.4 Intron XP_005273078.1
XM_005273022.3 Intron XP_005273079.1
XM_005273023.4 Intron XP_005273080.1
XM_006711727.3 Intron XP_006711790.1
XM_006711728.3 Intron XP_006711791.1
XM_011544021.2 Intron XP_011542323.1
XM_011544022.2 Intron XP_011542324.1
XM_011544023.2 Intron XP_011542325.1
XM_011544024.2 Intron XP_011542326.1
XM_011544026.2 Intron XP_011542328.1
XM_011544028.2 Intron XP_011542330.1
XM_011544029.2 Intron XP_011542331.1
XM_011544030.2 Intron XP_011542332.1
XM_017000103.1 Intron XP_016855592.1
XM_017000104.1 Intron XP_016855593.1
XM_017000105.1 Intron XP_016855594.1
XM_017000106.1 Intron XP_016855595.1
XM_017000107.1 Intron XP_016855596.1
XM_017000108.1 Intron XP_016855597.1
XM_017000109.1 Intron XP_016855598.1

View Full Product Details