Product Details

SNP ID
rs2336156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52839001 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGCCTCTGAGCTTGACAAACCACC[A/G]AGCAGTGCCCCTACCTTGCTGTTTC
Phenotype
MIM: 600564
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITIH4 PubMed Links

Gene Details

Gene
ITIH4
Gene Name
inter-alpha-trypsin inhibitor heavy chain family member 4
There are no transcripts associated with this gene.

Gene
MIR8064
Gene Name
microRNA 8064
There are no transcripts associated with this gene.

Gene
MUSTN1
Gene Name
musculoskeletal, embryonic nuclear protein 1
There are no transcripts associated with this gene.

Gene
TMEM110
Gene Name
transmembrane protein 110
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198563.2 2507 UTR 3 NP_940965.1
Gene
TMEM110-MUSTN1
Gene Name
TMEM110-MUSTN1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198974.2 2507 Intron NP_001185903.2

View Full Product Details