Product Details

SNP ID
rs2242084
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:3192269 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATGACGGCACCCCTGGAACTGTG[A/G]GTCCCCACAGCTGCAGTGGGTAAGG
Phenotype
MIM: 608998
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TSSC1 PubMed Links

Gene Details

Gene
TSSC1
Gene Name
tumor suppressing subtransferable candidate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006711893.2 Intron XP_006711956.1
XM_011510383.2 Intron XP_011508685.1
XM_011510384.2 Intron XP_011508686.1
XM_011510385.2 Intron XP_011508687.1
XM_011510387.2 Intron XP_011508689.1
XM_017004818.1 Intron XP_016860307.1

View Full Product Details