Product Details
- SNP ID
-
rs2446211
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:5783701 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTCCTTGGGGGGTGGGGGTGGCAG[C/T]GGGGCCGGGCCTTCCTCTAGGGGCG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CATSPERD
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs141874203] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CATSPERD
- Gene Name
- cation channel sperm associated auxiliary subunit delta
There are no transcripts associated with this gene.
- Gene
- DUS3L
- Gene Name
- dihydrouridine synthase 3 like
There are no transcripts associated with this gene.
- Gene
- PRR22
- Gene Name
- proline rich 22
View Full Product Details