Product Details

SNP ID
rs2636557
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184231762 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGTTCTTTCATTGATTCATTCA[C/T]ACATGGCTTTGATTCCTGCCCAGGG
Phenotype
MIM: 611620
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR1224 PubMed Links

Gene Details

Gene
MIR1224
Gene Name
microRNA 1224
There are no transcripts associated with this gene.

Gene
VWA5B2
Gene Name
von Willebrand factor A domain containing 5B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320373.1 Intron NP_001307302.1
NM_138345.2 Intron NP_612354.1
XM_011513288.2 Intron XP_011511590.1
XM_011513289.2 Intron XP_011511591.1
XM_011513290.2 Intron XP_011511592.1
XM_011513291.2 Intron XP_011511593.1
XM_011513292.2 Intron XP_011511594.1

View Full Product Details