Product Details
- SNP ID
-
rs2704055
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:31510771 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGAATTTTTTGGTTCATCTGAAA[A/G]CACTGCTTTGGATACAGGAACTAGC
- Phenotype
-
MIM: 125671
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DSG2
PubMed Links
Gene Details
- Gene
- DSG2
- Gene Name
- desmoglein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001943.4 |
|
Intron |
|
|
NP_001934.2 |
View Full Product Details