Product Details

SNP ID
rs2244012
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:132565533 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGTATACACTTTGACTTCTGGTGCT[A/G]GATCATGTCCTCATTCCCCGCCCTT
Phenotype
MIM: 147850 MIM: 604040
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL5 PubMed Links

Gene Details

Gene
IL5
Gene Name
interleukin 5
There are no transcripts associated with this gene.

Gene
RAD50
Gene Name
RAD50 double strand break repair protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005732.3 Intron NP_005723.2

View Full Product Details