Product Details

SNP ID
rs2737791
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48601524 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCCCTATGGTTCTCACACTGTA[G/C]GAGGACTGGCCAAGGGGCAGCATGA
Phenotype
MIM: 300512
Polymorphism
G/C, Transversion Substitution
Allele Nomenclature
Literature Links
WDR13 PubMed Links

Gene Details

Gene
WDR13
Gene Name
WD repeat domain 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166426.1 Intron NP_001159898.1
NM_017883.4 Intron NP_060353.2
XM_006724539.1 Intron XP_006724602.1
XM_006724541.3 Intron XP_006724604.1
XM_011543947.2 Intron XP_011542249.1

View Full Product Details