Product Details

SNP ID
rs3740318
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:75095568 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCCTCCACCATGCCTGATCCAGA[A/G]GTGCACCTTGGGGAACACTGAGGGC
Phenotype
MIM: 613191 MIM: 611575
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUSP13 PubMed Links

Gene Details

Gene
DUSP13
Gene Name
dual specificity phosphatase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007271.1 Intron NP_001007272.1
NM_001007272.1 Intron NP_001007273.1
NM_001007273.1 Intron NP_001007274.1
NM_001320842.1 Intron NP_001307771.1
NM_001320843.1 Intron NP_001307772.1
NM_016364.3 Intron NP_057448.3
XM_005269883.3 Intron XP_005269940.1
XM_005269884.4 Intron XP_005269941.3
XM_005269887.1 Intron XP_005269944.1
XM_005269890.1 Intron XP_005269947.1
XM_011539853.1 Intron XP_011538155.1
XM_011539854.2 Intron XP_011538156.1
XM_011539855.1 Intron XP_011538157.1
XM_011539856.2 Intron XP_011538158.1
XM_017016313.1 Intron XP_016871802.1
XM_017016314.1 Intron XP_016871803.1
Gene
SAMD8
Gene Name
sterile alpha motif domain containing 8
There are no transcripts associated with this gene.

View Full Product Details