Product Details

SNP ID
rs73343813
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:110881124 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAATATGTGACTGTAATTTACCTA[C/T]AAAAATGGTAATGTCATGTGATTCA
Phenotype
MIM: 608610
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDCD4 PubMed Links

Gene Details

Gene
PDCD4
Gene Name
programmed cell death 4 (neoplastic transformation inhibitor)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199492.1 Intron NP_001186421.1
NM_014456.4 Intron NP_055271.2
NM_145341.3 Intron NP_663314.1

View Full Product Details