Product Details

SNP ID
rs34358288
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:218285411 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAAAATGGTAACTGGAGCCGTAG[C/T]GTCGGTGCTGGAAGACGAGGCCACA
Phenotype
MIM: 611193
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RRP15 PubMed Links

Gene Details

Gene
RRP15
Gene Name
ribosomal RNA processing 15 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016052.3 125 Missense Mutation GCG,GTG A32V NP_057136.2
XM_011509597.2 125 Missense Mutation GCG,GTG A32V XP_011507899.1

View Full Product Details