Product Details
- SNP ID
-
hCV22274969
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:39343734 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTGGTCCATTTTTCTCCATCATT[C/G]TATACTCTCCTTTTTTTTCCCCCAC
- Phenotype
-
MIM: 606710
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LHFP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77539247] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LHFP
- Gene Name
- lipoma HMGIC fusion partner
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005780.2 |
1316 |
UTR 3 |
|
|
NP_005771.1 |
View Full Product Details