Product Details

SNP ID
rs3808976
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:45805799 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CGCGGGGTGACCCAGCTCCTCTGCT[G/A]CCATGAATAGGGCCCCTCTGAAGCG
Phenotype
MIM: 605881
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
SLC35C1 PubMed Links

Gene Details

Gene
SLC35C1
Gene Name
solute carrier family 35 member C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145265.1 710 Intron NP_001138737.1
NM_001145266.1 710 Intron NP_001138738.1
NM_018389.4 710 UTR 5 NP_060859.4
XM_011520202.2 710 Intron XP_011518504.1
XM_011520203.2 710 UTR 5 XP_011518505.1

View Full Product Details