Product Details

SNP ID
hCV25471983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29694886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCCTGGTTCCTCCCCGAGGGAT[T/C]GAAGCTGGTGCCGGTGGCCAGAGTG
Phenotype
MIM: 606248
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
QPRT PubMed Links

Gene Details

Gene
QPRT
Gene Name
quinolinate phosphoribosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318249.1 426 Intron NP_001305178.1
NM_001318250.1 426 Intron NP_001305179.1
NM_014298.4 426 Missense Mutation TCG,TTG S79L NP_055113.2

View Full Product Details