Product Details
- SNP ID
-
rs544296975
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:50354654 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCCATCGCCACCCCGGGGCTGCCG[C/T]TCGGGACCGGCAAGCCGGGTAGCCG
- Phenotype
-
MIM: 600610
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ10038
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113692002] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FLJ10038
- Gene Name
- uncharacterized protein FLJ10038
There are no transcripts associated with this gene.
- Gene
- GABPB1
- Gene Name
- GA binding protein transcription factor beta subunit 1
- Gene
- GABPB1-AS1
- Gene Name
- GABPB1 antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- MIR4712
- Gene Name
- microRNA 4712
There are no transcripts associated with this gene.
View Full Product Details