Product Details

SNP ID
rs11540720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40389572 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCAGGTACTTTATAGGTTTCTTTG[A/C]CCGTACAGATTTTGCCCGAGGAGCC
Phenotype
MIM: 126430
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TOP2A PubMed Links

Gene Details

Gene
TOP2A
Gene Name
topoisomerase (DNA) II alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001067.3 4644 Missense Mutation GCA,TCA A1515S NP_001058.2
XM_005257632.1 4644 Missense Mutation GCA,TCA A1503S XP_005257689.1
XM_011525165.1 4644 Intron XP_011523467.1

View Full Product Details