Product Details

SNP ID
rs35320927
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136496401 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCACCGCCAGGCTGCTGGGGCC[C/T]AGTGGCTGCACGTCTGCCTGGCTCG
Phenotype
MIM: 190198
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NOTCH1 PubMed Links

Gene Details

Gene
NOTCH1
Gene Name
notch 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017617.4 6630 Silent Mutation CTA,CTG L2446L NP_060087.3
XM_011518717.2 6630 Silent Mutation CTA,CTG L2205L XP_011517019.2

View Full Product Details