Product Details
- SNP ID
-
rs114896143
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:75603138 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAATTCTGCTTAGCAGCTTCTGGAA[C/T]GATCTGAAGAGTTCCACTTTGCTGA
- Phenotype
-
MIM: 604230
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ADAT1
PubMed Links
Gene Details
- Gene
- ADAT1
- Gene Name
- adenosine deaminase, tRNA specific 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001324444.1 |
2077 |
Silent Mutation |
TCA,TCG |
S292S |
NP_001311373.1 |
NM_001324445.1 |
2077 |
Silent Mutation |
TCA,TCG |
S441S |
NP_001311374.1 |
NM_001324446.1 |
2077 |
Silent Mutation |
TCA,TCG |
S292S |
NP_001311375.1 |
NM_001324448.1 |
2077 |
Silent Mutation |
TCA,TCG |
S359S |
NP_001311377.1 |
NM_001324449.1 |
2077 |
Silent Mutation |
TCA,TCG |
S359S |
NP_001311378.1 |
NM_001324450.1 |
2077 |
Silent Mutation |
TCA,TCG |
S210S |
NP_001311379.1 |
NM_001324451.1 |
2077 |
Silent Mutation |
TCA,TCG |
S210S |
NP_001311380.1 |
NM_001324452.1 |
2077 |
Silent Mutation |
TCA,TCG |
S354S |
NP_001311381.1 |
NM_001324453.1 |
2077 |
Silent Mutation |
TCA,TCG |
S272S |
NP_001311382.1 |
NM_012091.4 |
2077 |
Silent Mutation |
TCA,TCG |
S441S |
NP_036223.2 |
View Full Product Details