Product Details

SNP ID
rs746928955
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:63384206 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTCAGCTGCAGTGCCTCCCCAA[A/G]AGATCTGCTACCTTCATAGACAGAT
Phenotype
MIM: 601647
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PPP2R5E PubMed Links

Gene Details

Gene
PPP2R5E
Gene Name
protein phosphatase 2 regulatory subunit B'epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282179.1 Intron NP_001269108.1
NM_001282180.1 Intron NP_001269109.1
NM_001282181.1 Intron NP_001269110.1
NM_001282182.1 Intron NP_001269111.1
NM_006246.3 Intron NP_006237.1

View Full Product Details