Product Details

SNP ID
rs3764327
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31261808 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTTCAGGGATCCAGGTGAGACCCT[T/C]AGATGGAGCCCCCTTTCTCAGTGCT
Phenotype
MIM: 120980
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
ITGAM PubMed Links

Gene Details

Gene
ITGAM
Gene Name
integrin subunit alpha M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000632.3 Intron NP_000623.2
NM_001145808.1 Intron NP_001139280.1
XM_006721045.1 Intron XP_006721108.1
XM_011545850.2 Intron XP_011544152.1
XM_011545851.2 Intron XP_011544153.1
XM_017023216.1 Intron XP_016878705.1

View Full Product Details