Product Details

SNP ID
rs34351666
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:77697395 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCAACATGAACAACCGGAGGTCG[C/T]AGGATAACTTCAGGCCCTCCACCAT
Phenotype
MIM: 615146
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
USP33 PubMed Links

Gene Details

Gene
USP33
Gene Name
ubiquitin specific peptidase 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015017.4 3125 Silent Mutation CTA,CTG L917L NP_055832.3
NM_201624.2 3125 Silent Mutation CTA,CTG L886L NP_963918.1
NM_201626.2 3125 Intron NP_963920.1
XM_005270648.1 3125 Silent Mutation CTA,CTG L909L XP_005270705.1
XM_005270649.1 3125 UTR 3 XP_005270706.1
XM_011541055.1 3125 Silent Mutation CTA,CTG L818L XP_011539357.1
XM_011541056.2 3125 Intron XP_011539358.1
XM_017000722.1 3125 Silent Mutation CTA,CTG L878L XP_016856211.1
XM_017000723.1 3125 UTR 3 XP_016856212.1
XM_017000724.1 3125 UTR 3 XP_016856213.1
XM_017000725.1 3125 Silent Mutation CTA,CTG L787L XP_016856214.1
XM_017000726.1 3125 Silent Mutation CTA,CTG L779L XP_016856215.1
XM_017000727.1 3125 Silent Mutation CTA,CTG L738L XP_016856216.1
XM_017000728.1 3125 Silent Mutation CTA,CTG L738L XP_016856217.1
XM_017000729.1 3125 UTR 3 XP_016856218.1

View Full Product Details