Product Details

SNP ID
rs34255686
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132658339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGGAGCTCATTGAGGAGCTGGT[A/C]AACATCACCCAGAACCAGAAGGTGA
Phenotype
MIM: 147683
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IL13 PubMed Links

Gene Details

Gene
IL13
Gene Name
interleukin 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002188.2 167 Silent Mutation GTA,GTC V51V NP_002179.2
Gene
TH2LCRR
Gene Name
T helper type 2 locus control region associated RNA
There are no transcripts associated with this gene.

View Full Product Details