Product Details

SNP ID
rs35472059
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:102341487 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTATAGGTGCTGCTCTCTCATCC[C/T]GGGATGTCAGTGGGCTGCCTGTTTA
Phenotype
MIM: 613362
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CINP PubMed Links

Gene Details

Gene
CINP
Gene Name
cyclin dependent kinase 2 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320046.1 1957 Intron NP_001306975.1
NM_032630.2 1957 Intron NP_116019.1
Gene
ZNF839
Gene Name
zinc finger protein 839
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267827.1 1957 Missense Mutation CGG,TGG R582W NP_001254756.1
NM_001267828.1 1957 Missense Mutation CGG,TGG R582W NP_001254757.1
NM_018335.4 1957 Missense Mutation CGG,TGG R698W NP_060805.3
XM_006720203.3 1957 Missense Mutation CGG,TGG R610W XP_006720266.2
XM_011536945.1 1957 Missense Mutation CGG,TGG R732W XP_011535247.1
XM_011536946.2 1957 Missense Mutation CGG,TGG R682W XP_011535248.1
XM_011536948.2 1957 Missense Mutation CGG,TGG R582W XP_011535250.1
XM_011536949.2 1957 Intron XP_011535251.1
XM_011536950.2 1957 Intron XP_011535252.1
XM_017021447.1 1957 Missense Mutation CGG,TGG R648W XP_016876936.1
XM_017021448.1 1957 Intron XP_016876937.1
XM_017021449.1 1957 Missense Mutation CGG,TGG R363W XP_016876938.1
XM_017021450.1 1957 Intron XP_016876939.1
XM_017021451.1 1957 Intron XP_016876940.1

View Full Product Details