Product Details
- SNP ID
-
rs36053738
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
26
- Location
-
Chr.1:182575446 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCCACCAGCTCCATCACACTGAGG[C/T]TTGTTTGGACTGTGGGTTTGGGGGA
- Phenotype
-
MIM: 180435
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RNASEL
PubMed Links
Gene Details
- Gene
- RNASEL
- Gene Name
- ribonuclease L
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021133.3 |
2429 |
Silent Mutation |
AAA,AAG |
K724K |
NP_066956.1 |
View Full Product Details