Product Details

SNP ID
rs10838529
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:45909951 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTCGCTCCTATGGCTGCCGAGGC[A/G]AGCAGCTTCCCGGGCTCCATGAGGT
Phenotype
MIM: 604641 MIM: 603360
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C11orf94 PubMed Links

Gene Details

Gene
C11orf94
Gene Name
chromosome 11 open reading frame 94
There are no transcripts associated with this gene.

Gene
MAPK8IP1
Gene Name
mitogen-activated protein kinase 8 interacting protein 1
There are no transcripts associated with this gene.

Gene
PEX16
Gene Name
peroxisomal biogenesis factor 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004813.2 1626 UTR 3 NP_004804.1
NM_057174.2 1626 UTR 3 NP_476515.1
XM_011520474.1 1626 Intron XP_011518776.1

View Full Product Details