Product Details
- SNP ID
-
hCV25609627
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:29043360 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCAGGTACTGTGTCCCTTCTGCAC[C/T]CCCAGCTGTAAGGACTGTCCTACCC
- Phenotype
-
MIM: 616713
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PIPOX
PubMed Links
Gene Details
- Gene
- PIPOX
- Gene Name
- pipecolic acid and sarcosine oxidase
There are no transcripts associated with this gene.
View Full Product Details