Product Details
- SNP ID
-
rs7733544
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:127524453 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGGTAAGTTAAAAATTTTGCAAGAA[C/G]AAAGTAATTAGACAGTTTGAAAACT
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PRRC1
PubMed Links
Gene Details
- Gene
- PRRC1
- Gene Name
- proline rich coiled-coil 1
View Full Product Details