Product Details

SNP ID
rs74636418
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:109442176 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGATGGGCAGAAAGATGGTGGC[A/G]GTGCCCTGAGTTTCTATCTCCTCCT
Phenotype
MIM: 607129 MIM: 603498
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MICAL1 PubMed Links

Gene Details

Gene
MICAL1
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 1
There are no transcripts associated with this gene.

Gene
PPIL6
Gene Name
peptidylprolyl isomerase like 6
There are no transcripts associated with this gene.

Gene
SMPD2
Gene Name
sphingomyelin phosphodiesterase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003080.2 Intron NP_003071.2
XM_005267109.1 Intron XP_005267166.1
XM_011536079.1 Intron XP_011534381.1

View Full Product Details