Product Details
- SNP ID
-
hCV25615014
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:7174128 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCCAGCCAGCCTCCCAGACCCTCC[G/T]GGTCCTCACCTGCTCCTCCCAGGAC
- Phenotype
-
MIM: 108360
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ASGR1
PubMed Links
Gene Details
- Gene
- ASGR1
- Gene Name
- asialoglycoprotein receptor 1
View Full Product Details