Product Details

SNP ID
rs117020618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:171309820 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGGCGACCAGGCTCCAGGCCTC[C/T]GCCCTCTCGGGGCCAGAGGCGCCGC
Phenotype
MIM: 606141 MIM: 604640
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC728145 PubMed Links

Gene Details

Gene
LOC728145
Gene Name
collagen alpha-1(I) chain
There are no transcripts associated with this gene.

Gene
RANBP17
Gene Name
RAN binding protein 17
There are no transcripts associated with this gene.

Gene
TLX3
Gene Name
T-cell leukemia homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021025.3 Intron NP_066305.2

View Full Product Details