Product Details
- SNP ID
-
hCV25621713
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:63331672 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCCTGAAGAGCACTGGTCCAATCT[G/T]ACGTATTCGAAAACAATACATAAGT
- Phenotype
-
MIM: 136440
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KDSR
PubMed Links
Gene Details
- Gene
- KDSR
- Gene Name
- 3-ketodihydrosphingosine reductase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002035.2 |
1264 |
UTR 3 |
|
|
NP_002026.1 |
View Full Product Details