Product Details

SNP ID
rs34377353
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:126203652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCTTCTTCAGGGTGCCCACAGA[C/T]AGCTTCTATACAAAGAAAGGACAGA
Phenotype
MIM: 616587
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM118B PubMed Links

Gene Details

Gene
FAM118B
Gene Name
family with sequence similarity 118 member B
There are no transcripts associated with this gene.

Gene
RPUSD4
Gene Name
RNA pseudouridylate synthase domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144827.1 951 Silent Mutation CTA,CTG L269L NP_001138299.1
NM_032795.2 951 Silent Mutation CTA,CTG L300L NP_116184.2
XM_011543039.2 951 Silent Mutation CTA,CTG L107L XP_011541341.1

View Full Product Details