Product Details

SNP ID
rs3800990
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:141837741 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTTTGCCAGACCACTCAGGTAGC[A/G]TCTCTATTGGGTCTCCTTTCACTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRSS37 PubMed Links

Gene Details

Gene
PRSS37
Gene Name
protease, serine 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008270.2 Intron NP_001008271.2
NM_001171951.1 Intron NP_001165422.1
XM_005249946.4 Intron XP_005250003.1
XM_005249947.3 Intron XP_005250004.1
XM_011515792.2 Intron XP_011514094.1
XM_011515793.2 Intron XP_011514095.1

View Full Product Details