Product Details

SNP ID
rs11737783
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:6696937 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGTGGATAAATTGCTCAAGGACCT[A/G]GACGCCAATGGAGATGCCCAGGTGG
Phenotype
MIM: 600614
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC339988 PubMed Links

Gene Details

Gene
LOC339988
Gene Name
uncharacterized LOC339988
There are no transcripts associated with this gene.

Gene
S100P
Gene Name
S100 calcium binding protein P
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005980.2 277 Silent Mutation CTA,CTG L61L NP_005971.1

View Full Product Details