Product Details

SNP ID
rs150299337
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:31766760 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGAAGAATATAAAATCGTTTCT[G/T]ATTTTATATCAGCATCTACAGACAA
Phenotype
MIM: 614191
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DEPDC5 PubMed Links

Gene Details

Gene
DEPDC5
Gene Name
DEP domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007188.2 Intron NP_001007189.1
NM_001136029.2 Intron NP_001129501.1
NM_001242896.1 Intron NP_001229825.1
NM_001242897.1 Intron NP_001229826.1
NM_014662.4 Intron NP_055477.1
XM_005261862.1 Intron XP_005261919.1
XM_011530557.2 Intron XP_011528859.1
XM_011530558.2 Intron XP_011528860.1
XM_011530559.2 Intron XP_011528861.1
XM_011530560.2 Intron XP_011528862.1
XM_011530561.2 Intron XP_011528863.1
XM_011530562.2 Intron XP_011528864.1
XM_011530563.2 Intron XP_011528865.1
XM_011530565.2 Intron XP_011528867.1
XM_011530568.2 Intron XP_011528870.1
XM_011530569.2 Intron XP_011528871.1
XM_017029109.1 Intron XP_016884598.1
XM_017029110.1 Intron XP_016884599.1
XM_017029111.1 Intron XP_016884600.1
XM_017029112.1 Intron XP_016884601.1
XM_017029113.1 Intron XP_016884602.1
XM_017029114.1 Intron XP_016884603.1
XM_017029115.1 Intron XP_016884604.1

View Full Product Details