Product Details

SNP ID
rs145027704
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141524307 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATATCAAGCCCCACACAAAATG[A/C]ATGGCTATTGCTTGATTTCCCCTCT
Phenotype
MIM: 602121
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DIAPH1 PubMed Links

Gene Details

Gene
DIAPH1
Gene Name
diaphanous related formin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079812.2 Intron NP_001073280.1
NM_001314007.1 Intron NP_001300936.1
NM_005219.4 Intron NP_005210.3
XM_011537572.2 Intron XP_011535874.1
XM_011537573.2 Intron XP_011535875.1

View Full Product Details