Product Details

SNP ID
rs34174425
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:36497788 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTATATGGATATACCTGATATG[C/T]TCCCAGTCAACGCCTTCAAAAAAAG
Phenotype
MIM: 615932 MIM: 606964
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCTD20 PubMed Links

Gene Details

Gene
KCTD20
Gene Name
potassium channel tetramerization domain containing 20
There are no transcripts associated with this gene.

Gene
STK38
Gene Name
serine/threonine kinase 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305102.1 1493 Silent Mutation GAA,GAG E388E NP_001292031.1
NM_007271.3 1493 Silent Mutation GAA,GAG E388E NP_009202.1
XM_006714988.3 1493 Silent Mutation GAA,GAG E388E XP_006715051.1
XM_006714989.3 1493 Silent Mutation GAA,GAG E388E XP_006715052.1
XM_017010226.1 1493 Silent Mutation GAA,GAG E388E XP_016865715.1

View Full Product Details