Product Details

SNP ID
rs55756224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3067425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGAAGGCCCGAATGGTAATGCTC[C/T]TCCCTACTTCTGCTCAGGGGTTGGG
Phenotype
MIM: 606001 MIM: 608482
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL32 PubMed Links

Gene Details

Gene
IL32
Gene Name
interleukin 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012631.1 138 Intron NP_001012649.1
NM_001012632.1 138 Intron NP_001012650.1
NM_001012633.1 138 Intron NP_001012651.1
NM_001012634.1 138 Intron NP_001012652.1
NM_001012635.1 138 Intron NP_001012653.1
NM_001012636.1 138 Intron NP_001012654.1
NM_001012718.1 138 Intron NP_001012736.1
NM_001308078.1 138 Missense Mutation CTC,TTC L22F NP_001295007.1
NM_004221.4 138 Intron NP_004212.4
Gene
MMP25
Gene Name
matrix metallopeptidase 25
There are no transcripts associated with this gene.

Gene
MMP25-AS1
Gene Name
MMP25 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details