Product Details

SNP ID
rs61740898
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:209675694 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCCCAGAGCCGAGATGGAAACG[A/G]TCCAGGAGCTGATCCCCCTGGCCAA
Phenotype
MIM: 614447
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
G0S2 PubMed Links

Gene Details

Gene
G0S2
Gene Name
G0/G1 switch 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015714.3 267 Missense Mutation ATC,GTC I4V NP_056529.1
Gene
LOC101930114
Gene Name
uncharacterized LOC101930114
There are no transcripts associated with this gene.

View Full Product Details