Product Details

SNP ID
rs34080556
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:33585289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTTCATGACCTCCTCGTCCTCC[A/G]TATGAGGAATGTTCATAGCCACCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM98A PubMed Links

Gene Details

Gene
FAM98A
Gene Name
family with sequence similarity 98 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304538.1 1185 Silent Mutation TAC,TAT Y153Y NP_001291467.1
NM_015475.4 1185 Silent Mutation TAC,TAT Y348Y NP_056290.3

View Full Product Details