Product Details

SNP ID
rs6574978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:88011069 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTCCCTTTATGGATTGATTATAC[C/T]TGGAATAAAGACAACTGGACTTTCT
Phenotype
MIM: 604620
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR65 PubMed Links

Gene Details

Gene
GPR65
Gene Name
G protein-coupled receptor 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003608.3 780 Silent Mutation ACC,ACT T74T NP_003599.2
Gene
LOC101928791
Gene Name
uncharacterized LOC101928791
There are no transcripts associated with this gene.

View Full Product Details