Product Details

SNP ID
rs60448399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:118826081 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGACTCCGATGAGGATGTGATCCT[A/G]GTGGAGTCTCAGGACCCTCCACTTC
Phenotype
MIM: 300701
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZCCHC12 PubMed Links

Gene Details

Gene
ZCCHC12
Gene Name
zinc finger CCHC-type containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001312891.1 1339 Silent Mutation CTA,CTG L279L NP_001299820.1
NM_173798.3 1339 Silent Mutation CTA,CTG L279L NP_776159.1

View Full Product Details