Product Details
- SNP ID
-
rs8080510
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:63999336 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTAGTGCCTCCCCCACTGGACTTA[A/G]GGCTCCAGGAAGGTAGGCACCCTGG
- Phenotype
-
MIM: 146630
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ICAM2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80292971] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ICAM2
- Gene Name
- intercellular adhesion molecule 2
There are no transcripts associated with this gene.
- Gene
- PRR29
- Gene Name
- proline rich 29
- Gene
- PRR29-AS1
- Gene Name
- PRR29 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details