Product Details

SNP ID
rs61732444
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:111900452 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGCCTCATGCATGAAAATATCAC[A/G]AAAGAAAGTGGCTTCAGCAGCTGCA
Phenotype
MIM: 606723
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAM1A PubMed Links

Gene Details

Gene
ADAM1A
Gene Name
ADAM metallopeptidase domain 1A (pseudogene)
There are no transcripts associated with this gene.

Gene
MAPKAPK5
Gene Name
mitogen-activated protein kinase-activated protein kinase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003668.3 7399 Intron NP_003659.2
NM_139078.2 7399 Intron NP_620777.1
XM_017020125.1 7399 Intron XP_016875614.1
XM_017020126.1 7399 Intron XP_016875615.1
XM_017020127.1 7399 UTR 3 XP_016875616.1
XM_017020128.1 7399 UTR 3 XP_016875617.1
XM_017020129.1 7399 Intron XP_016875618.1
XM_017020130.1 7399 Intron XP_016875619.1
XM_017020131.1 7399 Intron XP_016875620.1
XM_017020132.1 7399 UTR 3 XP_016875621.1
XM_017020133.1 7399 UTR 3 XP_016875622.1
XM_017020134.1 7399 UTR 3 XP_016875623.1
XM_017020135.1 7399 UTR 3 XP_016875624.1
XM_017020136.1 7399 UTR 3 XP_016875625.1
XM_017020137.1 7399 Intron XP_016875626.1
XM_017020138.1 7399 UTR 3 XP_016875627.1
XM_017020139.1 7399 Intron XP_016875628.1
Gene
TMEM116
Gene Name
transmembrane protein 116
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193453.1 7399 Intron NP_001180382.1
NM_001193531.1 7399 Intron NP_001180460.1
NM_001294314.1 7399 Intron NP_001281243.1
NM_138341.2 7399 Intron NP_612350.1
XM_006719678.3 7399 Intron XP_006719741.1
XM_011538946.1 7399 Intron XP_011537248.1
XM_011538947.1 7399 Intron XP_011537249.1
XM_011538948.2 7399 Intron XP_011537250.1
XM_011538949.2 7399 Intron XP_011537251.1
XM_011538950.1 7399 Intron XP_011537252.1
XM_011538951.2 7399 Intron XP_011537253.1
XM_011538953.1 7399 Intron XP_011537255.1
XM_011538955.1 7399 Intron XP_011537257.1
XM_011538957.1 7399 Intron XP_011537259.1
XM_011538958.1 7399 Intron XP_011537260.1
XM_017020187.1 7399 Intron XP_016875676.1
XM_017020188.1 7399 Intron XP_016875677.1
XM_017020189.1 7399 Intron XP_016875678.1
XM_017020190.1 7399 Intron XP_016875679.1
XM_017020191.1 7399 Intron XP_016875680.1
XM_017020192.1 7399 Intron XP_016875681.1
XM_017020193.1 7399 Intron XP_016875682.1
XM_017020194.1 7399 Intron XP_016875683.1

View Full Product Details